5-Methyl-1H-pyrazole-3-carbonitrile 97%
Supplier: Ambeed
|
Warning
|
A354533-100MG
A354533-1G
A354533-250MG
A354533-5G
76978-536EA
26.53
USD
76978-536
76978-538
76978-540
76978-542
5-Methyl-1H-pyrazole-3-carbonitrile 97%
5-Methyl-1H-pyrazole-3-carbonitrile
Precaución: for Research Use Only
Formula:
C₅H₅N₃ |
MDL Number:
MFCD11934282 CAS Number: 38693-82-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...