Quinoline-2,4(1H,3H)-dione 97%

Supplier: Ambeed
Warning

A132202-10G A132202-1G A132202-25G A132202-5G
76972-138EA 75.33 USD
76972-138 76972-140 76972-142 76972-144
Quinoline-2,4(1H,3H)-dione 97%
Quinoline-2,4(1H,3H)-dione

Precaución: for Research Use Only

Formula: C₉H₇NO₂
MW: 161.15 g/mol
MDL Number: MFCD02937587
CAS Number: 52851-41-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR