2-(1,2-Benzisoxazol-3-yl)acetic acid 98%

Supplier: Ambeed
Warning

Synonyms: 1,2-Benzisoxazole-3-acetic Acid

A165286-100G A165286-25G A165286-5G
76973-758EA 147.47 USD
76973-758 76973-760 76973-762
2-(1,2-Benzisoxazol-3-yl)acetic acid 98%
2-(1,2-Benzisoxazol-3-yl)acetic acid

Precaución: for Research Use Only

Formula: C₉H₇NO₃
MW: 177.1568 g/mol
MDL Number: MFCD02180388
CAS Number: 4865-84-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR