4-(1-((tert-Butoxycarbonyl)amino)cyclobutyl)benzoic acid 95%

Supplier: Ambeed
Warning

A749877-100MG A749877-250MG
76981-360EA 264.17 USD
76981-360 76981-362
4-(1-((tert-Butoxycarbonyl)amino)cyclobutyl)benzoic acid 95%
4-(1-((tert-Butoxycarbonyl)amino)cyclobutyl)benzoic acid

Precaución: for Research Use Only

Formula: C₁₆H₂₁NO₄
MW: 291.34 g/mol
MDL Number: MFCD18762016
CAS Number: 1259223-99-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR