N4-Benzoyl-2'-deoxycytidine 98%

Supplier: Ambeed
Warning

Synonyms: N-(1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2-oxo-1,2-dihydropyrimidin-4-yl)benzamide

A914066-1G A914066-25G A914066-5G
76981-932EA 15.92 USD
76981-932 76981-934 76981-936
N4-Benzoyl-2'-deoxycytidine 98%
N4-Benzoyl-2'-deoxycytidine

Precaución: for Research Use Only

Formula: C₁₆H₁₇N₃O₅
MW: 331.33 g/mol
MDL Number: MFCD00010115
CAS Number: 4836-13-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR