(S)-1-(2,3-Difluorophenyl)ethanamine hydrochloride 95%

Supplier: Ambeed
Warning

A274615-100MG A274615-250MG
76977-258EA 43 USD
76977-258 76977-260
(S)-1-(2,3-Difluorophenyl)ethanamine hydrochloride 95%
(S)-1-(2,3-Difluorophenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₈H₁₀ClF₂N
MDL Number: MFCD12757704
CAS Number: 1415303-43-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR