4'-Bromobiphenyl-4-ylboronic acid 95%

Supplier: Ambeed
Warning

Synonyms: 4'-Bromo-4-biphenylboronic acid

A277986-10G A277986-1G A277986-250MG A277986-25G A277986-5G
76977-326EA 242.95 USD
76977-326 76977-328 76977-330 76977-332 76977-334
4'-Bromobiphenyl-4-ylboronic acid 95%
4'-Bromobiphenyl-4-ylboronic acid

Precaución: for Research Use Only

Formula: C₁₂H₁₀BBrO₂
MW: 276.9216 g/mol
MDL Number: MFCD02251652
CAS Number: 480996-05-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR