(2S,4S)-4-((tert-Butoxycarbonyl)amino)pyrrolidine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A531543-100MG A531543-1G A531543-250MG A531543-5G
76980-108EA 114.58 USD
76980-108 76980-110 76980-112 76980-114
(2S,4S)-4-((tert-Butoxycarbonyl)amino)pyrrolidine-2-carboxylic acid 98%
(2S,4S)-4-((tert-Butoxycarbonyl)amino)pyrrolidine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₀H₁₈N₂O₄
MW: 230.26 g/mol
MDL Number: MFCD08704525
CAS Number: 1279034-78-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR