2-Amino-4-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 5-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-4-(trifluoromethyl)pyridin-2-amine, 2-Amino-4-(trifluoromethyl)pyridine-5-boronic acid pinacol ester

A701998-100MG A701998-1G A701998-250MG
76981-098EA 45 USD
76981-098 76981-100 76981-102
2-Amino-4-(trifluoromethyl)-5-pyridineboronic acid pinacol ester 97%
2-Amino-4-(trifluoromethyl)-5-pyridineboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₂H₁₆BF₃N₂O₂
MW: 288.07 g/mol
MDL Number: MFCD12923420
CAS Number: 944401-57-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR