2-Cyano-5-nitrophenylboronic acid pinacol ester 97%

Supplier: Ambeed
Warning

Synonyms: 4-Nitro-2-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)benzonitrile

A600438-100MG A600438-1G A600438-250MG A600438-5G
76980-492EA 37 USD
76980-492 76980-494 76980-496 76980-498
2-Cyano-5-nitrophenylboronic acid pinacol ester 97%
2-Cyano-5-nitrophenylboronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₃H₁₅BN₂O₄
MDL Number: MFCD12546472
CAS Number: 1218791-28-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR