4-(Ethylamino)-3-nitrobenzeneboronic acid, pinacol ester 96%

Supplier: Ambeed
Warning

A531938-1G A531938-250MG A531938-5G
76980-116EA 152 USD
76980-116 76980-118 76980-120
4-(Ethylamino)-3-nitrobenzeneboronic acid, pinacol ester 96%
4-(Ethylamino)-3-nitrobenzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₂₁BN₂O₄
MDL Number: MFCD12026079
CAS Number: 1150271-70-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR