3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, pinacol ester 98%

Supplier: Ambeed
Warning

A525861-1G A525861-250MG A525861-5G
76980-042EA 24 USD
76980-042 76980-044 76980-046
3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, pinacol ester 98%
3-(Methoxycarbonyl)-5-nitrobenzeneboronic acid, pinacol ester

Precaución: for Research Use Only

Formula: C₁₄H₁₈BNO₆
MDL Number: MFCD09258774
CAS Number: 957061-12-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR