(3,5-Difluoro-2-nitrophenyl)boronic acid 96%

Supplier: Ambeed
Danger

A372555-100MG A372555-1G A372555-250MG
76978-716EA 39 USD
76978-716 76978-718 76978-720
(3,5-Difluoro-2-nitrophenyl)boronic acid 96%
(3,5-Difluoro-2-nitrophenyl)boronic acid

Precaución: for Research Use Only

Formula: C₆H₄BF₂NO₄
MDL Number: MFCD12025971
CAS Number: 1150114-60-5
UN: 2811
ADR: 6.1,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR