(R)-3-(Boc-amino)-3-(4-fluorophenyl)propionic acid 98+%

Supplier: Ambeed
Warning

Synonyms: (R)-3-((tert-Butoxycarbonyl)amino)-3-(4-fluorophenyl)propanoic acid

A469418-100MG A469418-1G A469418-250MG A469418-5G
76979-628EA 15.92 USD
76979-628 76979-630 76979-632 76979-634
(R)-3-(Boc-amino)-3-(4-fluorophenyl)propionic acid 98+%
(R)-3-(Boc-amino)-3-(4-fluorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₁₄H₁₈FNO₄
MW: 283.2954 g/mol
MDL Number: MFCD03427957
CAS Number: 479064-94-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR