(S)-tert-Butyl 2-methylpiperazine-1-carboxylate hydrochloride 95%
Supplier: Ambeed
|
Warning
|
A149133-10G
A149133-1G
A149133-250MG
A149133-5G
76973-028EA
79
USD
76973-028
76973-030
76973-032
76973-034
(S)-tert-Butyl 2-methylpiperazine-1-carboxylate hydrochloride 95%
(S)-tert-Butyl 2-methylpiperazine-1-carboxylate hydrochloride
Precaución: for Research Use Only
Formula:
C₁₀H₂₁ClN₂O₂ |
MDL Number:
MFCD11101258 CAS Number: 960283-58-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...