(R)-1-(4-Methoxyphenyl)ethanamine hydrochloride 98%

Supplier: Ambeed
Warning

A637508-1G A637508-5G A637508-250MG
76980-730EA 63 USD
76980-730 76980-732 77882-479
(R)-1-(4-Methoxyphenyl)ethanamine hydrochloride 98%
(R)-1-(4-Methoxyphenyl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₉H₁₄ClNO
MDL Number: MFCD12910566
CAS Number: 911373-69-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR