3-(3,5-Difluorophenyl)propionic acid 97%

Supplier: Ambeed
Warning

A553482-1G A553482-250MG A553482-5G A553482-25G
76980-238EA 18.04 USD
76980-238 76980-240 76980-242 77673-470
3-(3,5-Difluorophenyl)propionic acid 97%
3-(3,5-Difluorophenyl)propionic acid

Precaución: for Research Use Only

Formula: C₉H₈F₂O₂
MW: 186.16 g/mol
MDL Number: MFCD04116054
CAS Number: 84315-24-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR