N,N-Dimethyl 4-boronobenzenesulfonamide 95%

Supplier: Ambeed
Warning

Synonyms: 4-(N,N-Dimethylsulphamoyl)phenylboronic acid, 4-(Dimethylsulphamoyl)benzeneboronic acid

A648126-250MG A648126-5G
76980-786EA 23.34 USD
76980-786 76980-788
N,N-Dimethyl 4-boronobenzenesulfonamide 95%
N,N-Dimethyl 4-boronobenzenesulphonamide

Precaución: for Research Use Only

Formula: C₈H₁₂BNO₄S
MW: 229.06 g/mol
MDL Number: MFCD06659849
CAS Number: 486422-59-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR