(R)-tert-Butyl (2-aminopropyl)carbamate 97%

Supplier: Ambeed
Warning

A340304-100MG A340304-1G A340304-250MG A340304-25G A340304-5G
76978-386EA 16 USD
76978-386 76978-388 76978-390 76978-392 76978-394
(R)-tert-Butyl (2-aminopropyl)carbamate 97%
(R)-tert-Butyl (2-aminopropyl)carbamate

Precaución: for Research Use Only

Formula: C₈H₁₈N₂O₂
MDL Number: MFCD16295168
CAS Number: 333743-54-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR