(2R,4S)-4-Aminopyrrolidine-2-carboxylic acid dihydrochloride 97%
Supplier: Ambeed
|
Warning
|
A131755-100MG
A131755-1G
A131755-250MG
76972-112EA
131.56
USD
76972-112
76972-114
76972-116
(2R,4S)-4-Aminopyrrolidine-2-carboxylic acid dihydrochloride 97%
(2R,4S)-4-Aminopyrrolidine-2-carboxylic acid dihydrochloride
Precaución: for Research Use Only
Formula:
C₅H₁₂Cl₂N₂O₂ MW: 203.06 g/mol |
MDL Number:
MFCD09025424 CAS Number: 2829279-78-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...