Methyl 4-Benzyl-3-morpholinecarboxylate 95%

Supplier: Ambeed
Warning

A585875-1G A585875-250MG
76980-416EA 89.12 USD
76980-416 76980-418
Methyl 4-Benzyl-3-morpholinecarboxylate 95%
Methyl 4-Benzyl-3-morpholinecarboxylate

Precaución: for Research Use Only

Formula: C₁₃H₁₇NO₃
MW: 235.27 g/mol
MDL Number: MFCD09756515
CAS Number: 212650-44-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR