9,9-Bis(4-glycidyloxyphenyl)fluorene 95%

Supplier: Ambeed
Warning

Synonyms: 4,4'-(9-Fluorenylidene)bis(1,2-epoxy-3-phenoxypropane), 2,2'-((((9H-Fluorene-9,9-diyl)bis(4,1-phenylene))bis(oxy))bis(methylene))bis(oxirane)

A674616-100G A674616-25G A674616-5G
76980-948EA 132 USD
76980-948 76980-950 76980-952
9,9-Bis(4-glycidyloxyphenyl)fluorene 95%
9,9-Bis(4-glycidyloxyphenyl)fluorene

Precaución: for Research Use Only

Formula: C₃₁H₂₆O₄
MDL Number: MFCD02171148
CAS Number: 47758-37-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR