(4R,5S,6S)-(Pivaloyloxy)methyl 3-((1-(4,5-dihydrothiazol-2-yl)azetidin-3-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Tebipenem pivoxil

A259534-100MG A259534-10MG A259534-1MG A259534-25MG A259534-50MG A259534-5MG A259534-1G A259534-250MG
76976-890EA 22.28 USD
76976-890 76976-892 76976-894 76976-896 76976-898 76976-900 77612-484 77672-318
(4R,5S,6S)-(Pivaloyloxy)methyl 3-((1-(4,5-dihydrothiazol-2-yl)azetidin-3-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate 98%
(4R,5S,6S)-(Pivaloyloxy)methyl 3-((1-(4,5-dihydrothiazol-2-yl)azetidin-3-yl)thio)-6-((R)-1-hydroxyethyl)-4-methyl-7-oxo-1-azabicyclo[3.2.0]hept-2-ene-2-carboxylate

Precaución: for Research Use Only

Formula: C₂₂H₃₁N₃O₆S₂
MW: 497.62 g/mol
MDL Number: MFCD17215369
CAS Number: 161715-24-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR