(R)-Piperazin-2-ylmethanol dihydrochloride 97%

Supplier: Ambeed
Warning

A150465-100MG A150465-1G A150465-250MG
76973-078EA 22 USD
76973-078 76973-080 76973-082
(R)-Piperazin-2-ylmethanol dihydrochloride 97%
(R)-Piperazin-2-ylmethanol dihydrochloride

Precaución: for Research Use Only

Formula: C₅H₁₂N₂O·2ClH
MW: 189.083 g/mol
MDL Number: MFCD09954306
CAS Number: 1217482-26-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR