3-(Pyridin-2-yl)-1,2,4-thiadiazol-5-amine 97%

Supplier: Ambeed
Warning

A265774-100MG A265774-1G A265774-250MG A265774-5G
76977-114EA 23 USD
76977-114 76977-116 76977-118 77881-725
3-(Pyridin-2-yl)-1,2,4-thiadiazol-5-amine 97%
3-(Pyridin-2-yl)-1,2,4-thiadiazol-5-amine

Precaución: for Research Use Only

Formula: C₇H₆N₄S
MDL Number: MFCD11845692
CAS Number: 138588-22-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR