Methyl 5-oxazolecarboxylate 97%

Supplier: Ambeed
Warning

A232366-100MG A232366-250MG A232366-5G
76976-202EA 21.22 USD
76976-202 76976-204 76976-206
Methyl 5-oxazolecarboxylate 97%
Methyl 5-oxazolecarboxylate

Precaución: for Research Use Only

Formula: C₅H₅NO₃
MW: 127.09 g/mol
MDL Number: MFCD00144774
CAS Number: 121432-12-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR