6-Methylindole-3-carboxyaldehyde 98%

Supplier: Ambeed
Warning

Synonyms: 6-Methyl-1H-indole-3-carbaldehyde

A134188-10G A134188-1G A134188-25G A134188-5G
76972-260EA 127 USD
76972-260 76972-262 76972-264 76972-266
6-Methylindole-3-carboxyaldehyde 98%
6-Methylindole-3-carboxyaldehyde

Precaución: for Research Use Only

Formula: C₁₀H₉NO
MDL Number: MFCD00049347
CAS Number: 4771-49-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR