(2S,4S)-Methyl 4-(((benzyloxy)carbonyl)amino)pyrrolidine-2-carboxylate dihydrochloride 95%

Supplier: Ambeed
Warning

A554189-1G A554189-250MG
76980-246EA 211.12 USD
76980-246 76980-248
(2S,4S)-Methyl 4-(((benzyloxy)carbonyl)amino)pyrrolidine-2-carboxylate dihydrochloride 95%
(2S,4S)-Methyl 4-(((benzyloxy)carbonyl)amino)pyrrolidine-2-carboxylate dihydrochloride

Precaución: for Research Use Only

Formula: C₁₄H₂₀Cl₂N₂O₄
MW: 351.22 g/mol
MDL Number: MFCD11101179
CAS Number: 1279038-33-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR