Methyl-2-amino-4-oxazolecarboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 2-aminooxazole-4-carboxylate

A531241-1G A531241-25G A531241-5G
76980-088EA 40.32 USD
76980-088 76980-090 76980-092
Methyl-2-amino-4-oxazolecarboxylate 98%
Methyl 2-amino-4-oxazolecarboxylate

Precaución: for Research Use Only

Formula: C₅H₆N₂O₃
MW: 142.1127 g/mol
MDL Number: MFCD09870069
CAS Number: 1000576-38-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR