(6-Bromopyrazin-2-yl)methanol 95%

Supplier: Ambeed
Warning

A563031-100MG A563031-1G
76980-274EA 95 USD
76980-274 77673-506
(6-Bromopyrazin-2-yl)methanol 95%
(6-Bromopyrazin-2-yl)methanol

Precaución: for Research Use Only

Formula: C₅H₅BrN₂O
MDL Number: MFCD14702702
CAS Number: 1209458-06-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR