Methyl-1-methyl-1H-[1,2,4]triazole-3-carboxylate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 1-methyl-1H-[1,2,4]triazole-3-carboxylate

A282029-1G A282029-250MG A282029-25G A282029-5G
76977-442EA 53.05 USD
76977-442 76977-444 77593-740 77672-456
Methyl-1-methyl-1H-[1,2,4]triazole-3-carboxylate 98%
Methyl-1-methyl-1H-[1,2,4]triazole-3-carboxylate

Precaución: for Research Use Only

Formula: C₅H₇N₃O₂
MW: 141.1300 g/mol
MDL Number: MFCD13191558
CAS Number: 57031-66-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR