Sodium ((2R,3S,4R,5R)-3,4-dihydroxy-5-(6-hydroxy-9H-purin-9-yl)tetrahydrofuran-2-yl)methyl phosphate, (∼25% H₂O) 99% + (by HPLC) (contains < 25% H₂O)

Supplier: Ambeed
Warning

A666161-100G A666161-25G A666161-500G
76980-892EA 35.01 USD
76980-892 76980-894 76980-896
Sodium ((2R,3S,4R,5R)-3,4-dihydroxy-5-(6-hydroxy-9H-purin-9-yl)tetrahydrofuran-2-yl)methyl phosphate, (∼25% H₂O) 99% + (by HPLC) (contains < 25% H₂O)
Sodium ((2R,3S,4R,5R)-3,4-dihydroxy-5-(6-hydroxy-9H-purin-9-yl)tetrahydrofuran-2-yl)methyl phosphate

Precaución: for Research Use Only

Formula: C₁₀H₁₁N₄Na₂O₈P
MW: 392.16 g/mol
MDL Number: MFCD00036201
CAS Number: 4691-65-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR