1-Acetyl-4-piperidineacetic acid 98%

Supplier: Ambeed
Warning

A226905-1G A226905-250MG A226905-5G
76975-976EA 37.14 USD
76975-976 76975-978 76975-980
1-Acetyl-4-piperidineacetic acid 98%
1-Acetyl-4-piperidineacetic acid

Precaución: for Research Use Only

Formula: C₉H₁₅NO₃
MW: 185.22 g/mol
MDL Number: MFCD12027293
CAS Number: 78056-60-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR