Sodium (S)-6-methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulfinyl)benzo[d]imidazol-1-ide 98%

Supplier: Ambeed
Danger

A188600-100G A188600-1G A188600-250MG A188600-25G A188600-5G A188600-10G
76974-764EA 228 USD
76974-764 76974-766 76974-768 76974-770 76974-772 77881-406
Sodium (S)-6-methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulfinyl)benzo[d]imidazol-1-ide 98%
Sodium (S)-6-methoxy-2-(((4-methoxy-3,5-dimethylpyridin-2-yl)methyl)sulfinyl)benzo[d]imidazol-1-ide

Precaución: for Research Use Only

Formula: C₁₇H₁₈N₃NaO₃S
MDL Number: MFCD07802816
CAS Number: 161796-78-7
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR