2-Amino-5-bromo-6-fluorobenzonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 6-Amino-3-bromo-2-fluorobenzonitrile

A183272-1G A183272-250MG A183272-25G A183272-5G
76974-516EA 9.55 USD
76974-516 76974-518 76974-520 76974-522
2-Amino-5-bromo-6-fluorobenzonitrile 97%
2-Amino-5-bromo-6-fluorobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₄BrFN₂
MW: 215.0225 g/mol
MDL Number: MFCD06657979
CAS Number: 845866-92-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR