1-Boc-1H-pyrazole-5-boronic acid 95%

Supplier: Ambeed
Warning

Synonyms: (1-(tert-Butoxycarbonyl)-1H-pyrazol-5-yl)boronic acid, 1H-Pyrazole-5-boronic acid, N1-BOC protected, 1-Boc-1H-pyrazole-5-boronic Acid

A621944-1G A621944-250MG A621944-25G A621944-5G
76980-628EA 88.06 USD
76980-628 76980-630 76980-632 76980-634
1-Boc-1H-pyrazole-5-boronic acid 95%
1-Boc-1H-pyrazole-5-boronic acid

Precaución: for Research Use Only

Formula: C₈H₁₃BN₂O₄
MW: 212.0108 g/mol
MDL Number: MFCD12025961
CAS Number: 1217500-54-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR