1-Chloro-4-(trifluoromethoxy)benzene 98%

Supplier: Ambeed
Warning

A123082-100G A123082-5G
76971-658EA 33 USD
76971-658 76971-660
1-Chloro-4-(trifluoromethoxy)benzene 98%
1-Chloro-4-(trifluoromethoxy)benzene

Precaución: for Research Use Only

Formula: C₇H₄ClF₃O
MDL Number: MFCD00040953
CAS Number: 461-81-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR