2-Methyl-6-phenylpyridine 98%

Supplier: Ambeed
Warning

A791513-100MG A791513-1G A791513-250MG
76981-622EA 44 USD
76981-622 76981-624 76981-626
2-Methyl-6-phenylpyridine 98%
2-Methyl-6-phenylpyridine

Precaución: for Research Use Only

Formula: C₁₂H₁₁N
MDL Number: MFCD00796537
CAS Number: 46181-30-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR