Methyl-4-amino-2-chlorobenzoate 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Methyl 4-amino-2-chlorobenzoate
A156126-100G
A156126-10G
A156126-1G
A156126-25G
A156126-5G
76973-312EA
265.23
USD
76973-312
76973-314
76973-316
76973-318
76973-320
Methyl-4-amino-2-chlorobenzoate 97%
Methyl 4-amino-2-chlorobenzoate
Precaución: for Research Use Only
Formula:
C₈H₈ClNO₂ MW: 185.6076 g/mol |
MDL Number:
MFCD03840484 CAS Number: 46004-37-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...