Ethyl 3-bromoisonicotinate 98%
Supplier: Ambeed
|
Warning
|
A627507-1G
A627507-250MG
A627507-5G
76980-658EA
58
USD
76980-658
76980-660
77882-459
Ethyl 3-bromoisonicotinate 98%
Ethyl 3-bromoisonicotinate
Precaución: for Research Use Only
Formula:
C₈H₈BrNO₂ |
MDL Number:
MFCD11226640 CAS Number: 13959-01-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...