N4-Methylpyridine-3,4-diamine 95%

Supplier: Ambeed
Warning

A113947-100MG A113947-1G A113947-250MG A113947-5G
76971-050EA 20 USD
76971-050 76971-052 76971-054 77670-596
N4-Methylpyridine-3,4-diamine 95%
N4-Methylpyridine-3,4-diamine

Precaución: for Research Use Only

Formula: C₆H₉N₃
MDL Number: MFCD00982623
CAS Number: 1839-17-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR