(4-Fluorophenyl)acetonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 4-Fluorobenzyl cyanide, 4-Fluorophenylacetonitrile

A195648-100G A195648-10G A195648-25G A195648-500G A195648-5G
76975-018EA 26.53 USD
76975-018 76975-020 76975-022 76975-024 76975-026
(4-Fluorophenyl)acetonitrile 97%
(4-Fluorophenyl)acetonitrile

Precaución: for Research Use Only

Formula: C₈H₆FN
MW: 135.14 g/mol
MDL Number: MFCD00001917
CAS Number: 459-22-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR