3,4-Dibromo-5-fluoropyridine 95%

Supplier: Ambeed
Warning

A179968-100MG A179968-1G A179968-250MG A179968-5G
76974-416EA 16 USD
76974-416 76974-418 76974-420 76974-422
3,4-Dibromo-5-fluoropyridine 95%
3,4-Dibromo-5-fluoropyridine

Precaución: for Research Use Only

Formula: C5H2Br2FN
MDL Number: MFCD13193300
CAS Number: 1260843-59-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR