3-Fluoro-5-methylanisole 98%

Supplier: Ambeed
Warning

A117469-10G A117469-1G A117469-5G
76971-288EA 179 USD
76971-288 76971-290 76971-292
3-Fluoro-5-methylanisole 98%
3-Fluoro-5-methylanisole

Precaución: for Research Use Only

Formula: C₈H₉FO
MDL Number: MFCD13185608
CAS Number: 160911-11-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR