Methyl 2,4-dihydroxy-6-methylpyrimidine-5-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A752109-1G
A752109-250MG
76981-372EA
389.36
USD
76981-372
76981-374
Methyl 2,4-dihydroxy-6-methylpyrimidine-5-carboxylate 95%
Methyl 2,4-dihydroxy-6-methylpyrimidine-5-carboxylate
Precaución: for Research Use Only
Formula:
C₇H₈N₂O₄ MW: 184.15 g/mol |
MDL Number:
MFCD20133912 CAS Number: 869891-41-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...