Benzyl-2,4-dichloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate 95%

Supplier: Ambeed
Warning

Synonyms: Benzyl 2,4-dichloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate

A147466-1G A147466-250MG A147466-5G
76972-902EA 93.36 USD
76972-902 76972-904 76972-906
Benzyl-2,4-dichloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate 95%
Benzyl-2,4-dichloro-5,6-dihydropyrido[3,4-d]pyrimidine-7(8H)-carboxylate

Precaución: for Research Use Only

Formula: C₁₅H₁₃Cl₂N₃O₂
MW: 338.1886 g/mol
MDL Number: MFCD19707615
CAS Number: 1370411-44-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR