Methyl-5-fluoronicotinate 97%

Supplier: Ambeed
Warning

Synonyms: 5-Fluoro-nicotinic acid methyl ester, Methyl 5-fluoro-3-pyridinecarboxylate

A131234-100G A131234-100MG A131234-10G A131234-1G A131234-250MG A131234-25G A131234-5G
76972-070EA 464.68 USD
76972-070 76972-072 76972-074 76972-076 76972-078 76972-080 76972-082
Methyl-5-fluoronicotinate 97%
Methyl-5-fluoronicotinate

Precaución: for Research Use Only

Formula: C₇H₆FNO₂
MW: 155.13 g/mol
MDL Number: MFCD01692049
CAS Number: 455-70-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR