Adenosine 5'-monophosphate disodium salt (AMP disodium salt) dihydrate 95%
Supplier: Ambeed
|
Warning
|
A141922-100G
A141922-100MG
76972-644EA
5.31
USD
76972-644
77671-024
77671-026
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) dihydrate 95%
Adenosine 5'-monophosphate disodium salt (AMP disodium salt) dihydrate
Precaución: for Research Use Only
Formula:
C₁₀H₁₂N₅Na₂O₇P·2H₂O |
MDL Number:
MFCD00065023 CAS Number: 4578-31-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...