α-Bromo-2-chloro-4-fluorotoluene 98%

Supplier: Ambeed
Danger

A245243-100G A245243-5G A245243-25G
76976-570EA 76 USD
76976-570 76976-572 77550-574
α-Bromo-2-chloro-4-fluorotoluene 98%
α-Bromo-2-chloro-4-fluorotoluene

Precaución: for Research Use Only

Formula: C₇H₅BrClF
MDL Number: MFCD00236025
CAS Number: 45767-66-6
UN: 3261
ADR: 8,Ⅱ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR