2-Pyridinesulfonamide 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
Pyridine-2-sulphonamide
A168147-100MG
A168147-1G
A168147-250MG
A168147-5G
76973-876EA
14.86
USD
76973-876
76973-878
76973-880
76973-882
2-Pyridinesulfonamide 97%
2-Pyridinesulfonamide
Precaución: for Research Use Only
Formula:
C₅H₆N₂O₂S MW: 158.18 g/mol |
MDL Number:
MFCD09260336 CAS Number: 63636-89-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...